Nalgene® Utility Funnels, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Useful in numerous laboratory applications
Funnel 30255-383 has a large throat opening for faster transfer of materials.
The three smaller sizes are molded of autoclavable polypropylene; the three larger sizes are high-density polyethylene.
Customers Who Bought This Also Bought