519 Value Over Spectacle
# de catalogue 77393-736
Fournisseur: UNIVET OPTICAL TECHNOLOGIES
New Product
Spécifications
- Lens Coating:Scratch-resistant/hardcoat
- Lens Tint:Clear
- Frame Color:Black
Spécifications
A propos de cet article
The 519 Value over spectacle can be worn alone or over prescription glasses.
- Anti-scratch lens treatment
- Temples are adjustable in length and inclination
- Side venting
Over-molded terminal provides better comfort and stability.
Customers Who Bought This Also Bought