6×3 X-Gen Goggles
Fournisseur: UNIVET OPTICAL TECHNOLOGIES
New Product
Certificats
A propos de cet article
6×3 X-Gen goggles feature a panoramic lens that easily integrates with prescription glasses.
- Lens treatment: anti-fog and anti-scratch (double coating)
- Interchangeable lens system
- Frame engineered for soft facial support
- Compatible with filtering half masks
6×3 X-Gen goggles feature a lens with anti-fog and anti-scratch ultra performing Vanguard UDC treatment. They are certified according to EN166 standard, level 3 and 4 (EU) for protection against droplets, splashes, and large dust particles.
Ordering information: The 6×3 goggle face shield is available separately as a face protection accessory for this goggle series.
Customers Who Bought This Also Bought