5×3 X-Gen Spectacles
Fournisseur: UNIVET OPTICAL TECHNOLOGIES
New Product
Certificats
A propos de cet article
The 5×3 spectacles feature floating lens frame technology, which adapts to face conformation with no need for adjustments.
- Ultra-wraparound lens for better protection around the eyes
- Multiple lens treatments are available, including anti-fog, anti-scratch, and UV525
Designed to avoid annoying contact points with the face, these spectacles feature a frame with Floating Lens technology, available with anti-fog and anti-scratch ultra performing vanguard ultra (UDC) treatment. UV525 treatment is also available to protect eyes during curing light usage.
Customers Who Bought This Also Bought