Disposable Lens Eye Protection Kit, 712
Fournisseur: UNIVET OPTICAL TECHNOLOGIES
New Product
Certificats
A propos de cet article
This kit contains five frames (one each of blue, green, yellow, magenta, and black) and 20 interchangeable lenses.
- Anti-fog lens treatment
- Frame colors: blue, green, yellow, magenta, black
- Clear lenses
- Kit contains 5 frames/20 interchangeable lenses
Disposable eyewear features an interchangeable lens, and an ultra-lightweight design for comfortable extended use.
Ordering information: Replacement anti-fog lenses for the 712 eye protectors are available separately.
Customers Who Bought This Also Bought