VWR® Incineration Bags
Supplier: VWR International
Certificates
About this item
Constructed of durable polypropylene that meets the 165 g dart drop standard performed per ASTM D1709-98 and the 480 g tear standard performed per ASTM D1299-94A.
- Durable polypropylene construction
- Not autoclavable
- Marked with biohazard symbol
Red bags feature the universal biohazard symbol in black.
Note: Autoclaving not recommended. Bags will melt at 98.9 °C (210 °F).
Customers Who Bought This Also Bought