SP Bel-Art 4-Color Wash Bottles, Right-to-Know and Safety-Labeled, Wide Mouth, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Right-to-Know safety-labeled wash bottles feature a 53 mm (2") wide mouth for easy refilling.
- Pre-printed with chemical name and formula
- Leakproof
- Translucent LDPE construction
- Color-coded polypropylene closures for quick identification
Bottles are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
*Note: Use LYOB only with solvents compatible with LDPE.
Leak-proof, these translucent* LDPE bottles have color-coded polypropylene closures to aid in quick and easy identification.
Certifications: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Ordering information: Bottles are packaged 4/bag. The Assortment item includes 6/bag, with one each of Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and Label Your Own Bottle (LYOB).
Customers Who Bought This Also Bought