Sterilizable Overspec Goggles, 619 Polycarbonate
Fournisseur: UNIVET OPTICAL TECHNOLOGIES
New Product
Certificats
A propos de cet article
619 Polycarbonate sterilizable overspec goggles are available in direct vent or indirect vent formats.
- Overspec goggles feature interchangeable lens technology
- Meets EN 166 and EN 170 standards
- Scale number: (2C-1.2)
- Mechanical resistance: Medium energy impact (B)
- Optical class (1)
Goggles with polycarbonate lens are reusable for up to 30 autoclave sterilization cycles with steam at 121 °C, 1 atm for 30 minutes. They are also reusable for up to 10 gamma irradiation cycles.
Customers Who Bought This Also Bought