Sterilizable Goggles, 611 Polycarbonate AF121
Fournisseur: UNIVET OPTICAL TECHNOLOGIES
New Product
Certificats
A propos de cet article
611 Polycarbonate AF121 sterilizable goggles are available in direct vent or indirect vent formats. Both formats feature an anti-fog and anti-scratch lens treatment.
- Goggles feature interchangeable lens technology
- Meets EN 166 and EN 170 standards
- Scale number: (2C-1.2)
- Mechanical resistance: Medium energy impact (B)
- Optical class (1)
Goggles with polycarbonate lens are reusable for up to 40 autoclave sterilization cycles with steam at 121 °C, 1 atm for 30 minutes. The AF121 treatment applied to the lens prevents scratches and fogging at the same time. Goggles will withstand up to 10 steam sterilization cycles. They are also reusable for up to 10 gamma irradiation cycles.
Customers Who Bought This Also Bought