Sterilizable Goggles, 611 Byoxene
Fournisseur: UNIVET OPTICAL TECHNOLOGIES
New Product
Certificats
A propos de cet article
611 Byoxene sterilizable goggles are available in direct vent or indirect vent formats.
- Lens stays clear thanks to Byoxene lens technology
- Byoxene lenses cannot be autoclaved
- Goggles feature interchangeable lens technology
- Meets EN 166 standards
- Mechanical resistance: Medium energy impact (B)
- Optical class (1)
Goggles with Byoxene technology maintain lens transparency for up to 20 cycles of gamma sterilization (@ 25 kGy). The Byoxene lenses cannot be autoclaved.
Customers Who Bought This Also Bought