Nalgene® Wash Bottles, Teflon® FEP, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These transparent, chemical-resistant wash bottles are molded entirely of fluoropolymers to provide contamination-free dispensing.
Rinse with or dispense strong solvents or other liquids that might damage other plastics. One-piece Tefzel® polymer stem closure and tubing for leakproof service. Autoclavable. Packaged individually.
Frequently Bought Together