Nalgene® Wash Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Wash bottles feature one-piece polypropylene stem and closure in distinctive blue.
The transparent design makes liquid contents easy to view. Bottle is flexible for easy dispensing. Draw tube is welded into cap to prevent separation. Nalgene® catalog number and size code are printed on the side. Leakproof. Autoclavable.
Frequently Bought Together