Nalgene® Wash Bottles, Low-Density Polyethylene, Wide Mouth, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Wide mouth design for easy filling.
Through-the-top polypropylene screw closure and stem molded in one piece. Tip can be cut back to increase flow. Leakproof.
Caution: Not recommended for long-term use with acetone.
Customers Who Bought This Also Bought