ContactUsLinkComponent
DuraMaster® Cow Grain Leather Palm Gloves, Magid
DuraMaster® Cow Grain Leather Palm Gloves, Magid
# de catalogue 69100-110
Fournisseur:  Magid Glove
Small Business Enterprise
Grain Leather Palm Glove, Extra-Large
DuraMaster® Cow Grain Leather Palm Gloves, Magid
# de catalogue 69100-110
Fournisseur:  Magid Glove
Small Business Enterprise
Spécifications
  • Ambidextrous/Hand-specific:
    Ambidextrous
  • Glove Color:
    Brown
  • Glove size:
    XL
  • Material:
    Leather
  • Cat. no.:
    69100-110
  • Supplier No.:
    TB525EXL
Spécifications
A propos de cet article
The Magid® full leather glove features a gunn cut pattern, wing thumb and a select grain leather palm to deliver comfortable, long lasting wear. Grain leather is durable, cost-effective and is superior to split leather in repelling liquid in wet or outdoor applications.
  • Gunn cut pattern with split leather palm, wing thumb and brown denim back
  • Full leather index fingers and leather tips
  • Knuckle strap
  • Pull patch
  • Shirred elastic
This dependable glove also features full leather index finger and tips, a knuckle strap and 2.5" PE safety cuff for protection where it's needed most. The glove's pull patch helps to prevent the cuff from becoming detached during removal.
Grain work gloves sports blue striped denim back with elastic at the wrist for a secure and comfortable fit that breathes and helps wick moisture away from the hand.
Customers Who Bought This Also Bought