VWR® Adjustable Spray Wash Bottle
# de catalogue 16650-049
Fournisseur: VWR International
Spécifications
- Capacity:1 L (33.8 oz.)
- Cat. no.:16650-049
Spécifications
A propos de cet article
This double-headed wash bottle has an adjustable-spray stream and will keep contents completely free of contaminants if the sprayer and side-fill opening are in the closed position.
- Convenient double-headed design and side-fill opening
- Instant-off release eliminates drips
- High-density polyethylene construction
The convenient side-fill opening helps to maintain the purity of solutions. Large capacity minimizes refilling. Adjustable spray pattern is controlled by a contoured spray trigger with 'instant off.'Virtually closed system eliminates drips and handles hot, cold, aqueous, and solvent-based solutions. Engraved markings show liquid amount at a glance. High-density polyethylene bottle with polypropylene sprayer.
Customers Who Bought This Also Bought