About this item
MechPro plus gloves have all the features of MechPro standard gloves and additional padding in the palm area to reduce bruising and damage from vibration and excessive grip pressure.
- Padding minimizes Ssock
- Heat-resistant knit fabric
- Reinforced fingertips
Padding helps to minimize shock, which can lead to carpal tunnel syndrome.
Gusseted finger crotches provide additional comfort and flexibility. Gloves are double-stitched in high wear areas: fingertips, index fingers, and palms. An elastic cuff with hook and loop closure provides a secure fit that keeps out debris.
Heavy-duty, thick-ribbed, knit fabric back provides protection from nicks and abrasions and resists incidental heat. A synthetic leather palm and fingertips also offer protection while enhancing grip. Reinforced fingertips feature pinched seams to increase tactile sensitivity.
Specifications
- Catalog No:
- 83007-986
- 83007-990
- 83007-988
- 83007-994
- 83007-992
- 83007-998
- 83007-996
- 83008-002
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous