VWR® Reclosable Pink Anti-Static Bags
Fournisseur: VWR International
Certificats
A propos de cet article
These flexible and electrostatic-free 4 mm bags qualify as Type II entitled barrier material under MIL-B-81750b.
- Amine-free
- Reclosable design
- Pink tint for easy identification
They dissipate an induced 5000 V charge to technical zero in accordance with FTS 101C, Method 4046.1. Bags protect contents from damage caused by static electricity, in addition to providing protection against dust and moisture.
Customers Who Bought This Also Bought