Ultraclean* Cleanroom Bags and Sheeting, Polyethylene, KNF Cleanroom Products
Supplier: KNF Cleanroom Products
Certificates
About this item
This low-density polyethylene film is certified to Level II cleanliness standards.
Puncture and chemical-resistant, low vapor transmission rate, and good electrical properties. Can be easily heat sealed.
Customers Who Bought This Also Bought