HyFlex® 70-400 Cut Resistant Gloves, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
Type 70-400 cut-resistant gloves are knit from 100% Kevlar® continuous filament fiber.
- Gloves are produced in a silicone-free environment
- They have been prewashed to prevent shrinkage
These gloves are suitable for use in computer manufacturing, electronics and electrical assembly, and automotive paint room applications
They can also be used as cleanroom undergloves.
Customers Who Bought This Also Bought