Macron Fine Chemicals™ StandARd®, Platinum Cobalt Color Standard (APHA 500), StandARd®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Corrosive
APHA 500. Contains 0.13% chloroplatinic acid, 0.11% cobalt (II) chloride, and 4.6% hydrogen chloride in water.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.