KIMAX® Heavy-Duty Solution Bottles, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Borosilicate glass carboy-style bottles feature a narrow neck and sloping shoulders.
Necks are tooled for a uniform fit with a #12 rubber stopper (not included). The 19 L (5 gal.) size conforms to Federal Specification DD-B-597.
Customers Who Bought This Also Bought