Brady® Battery with Pistol Grip for HH83, HH85, EXA81 Readers
# de catalogue 77463-544
Fournisseur: Brady Worldwide
New Product
Spécifications
- Description:Lithium-ion battery with pistol grip
- Cat. no.:77463-544
Spécifications
A propos de cet article
Keep productivity high and workflow interruptions to a minimum with this long-lasting battery for the EXA81 and HH series devices.
- For use with HH83, HH85, EXA81 readers
- Color: Blue
- Dimensions: 6.5W×2.4"D
This 10050 mAh battery provides up to 18 hours of operation and features a pistol grip with trigger button. Direct back-up or replacement for HH85 or EXA81 devices; use with HH83 to add pistol grip with trigger functionality and expand battery life. Battery can he charged as a stand-alone unit in charging dock (not included), or when installed in EXA81 and HH series devices.
Customers Who Bought This Also Bought