VWR® REDISHIP Polyurethane Laboratory Chairs
Fournisseur: VWR International
New Product
Certificats
A propos de cet article
VWR® polyurethane seating is perfect for use in almost any environment with its enhanced durability and functionality.
- Rugged and durable
- Chemical-resistant
- Non-porous
- Easy to clean
- Convenient adjustability
- Ships assembled (requires only backbar insertion)
- Equipped with both glides and casters
- Weight capacity: 300 lbs.
- American made
- 15 year warranty
VWR® polyurethane seating features ergonomic adjustments for seat height and locking back tilt, a nylon-reinforced fiberglass base shipped with both dual-wheel, free-rolling casters and stationary glides. Three different height ranges are available for the chairs and can be selected with either a black or chrome footring on medium and high bench height chairs.
Certifications: Meets or exceeds ANSI/BIFMA industry standards.
Ordering information: All chairs come with a 15-year parts warranty and lifetime cylinder warranty.
Delivery: Both stationary glides and free-rolling casters are included with purchase. All product variations are ready for quick delivery through the REDISHIP program.
Packaging: Chairs are shipped assembled and will only require the back bar to be inserted into the chair base.
Spécifications
- Catalog No:
- 77449-176
- 77449-178
- 77449-180
- 77449-182
- 77449-184
- Seat/back material:
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Weight capacity:
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.