BIOXP® 3250 NGS Kits for Genomes
Fournisseur: TELESIS BIO
New Product
Certificats
A propos de cet article
Generate Illumina-compatible, adapter-ligated, NGS libraries for whole genome sequencing in a simple automated workflow using the BioXp 3250.
- Input: 10 to 20 ng of genomic DNA
- Compatible with Illumina sequencers
- 15 minutes of hands on time
- Compatible with BioXp 9600
The BioXp NGS library prep kit for whole genome sequencing provides the necessary reagents, when combined with customer-provided sequencing adapters, to generate Illumina-compatible NGS libraries for assessment of genomic DNA. This kit uses a streamlined, fully automated ligation-based library prep method. The input into the kit is 10 to 20 ng of purified plasmids in nuclease-free water, Tris-HCl pH 8.0, or Tris low EDTA (0.1 mM).
Delivery: Not supplied with necessary sequencing adapters.
Customers Who Bought This Also Bought