Stretch Nylon Gloves, Protective Industrial Products
Supplier: PIP
Certificates
About this item
Lint-free gloves are composed of 100% stretch nylon material that provides limited heat protection.
- Material is not treated with chemicals of any kind
- Heavyweight fabric is approximately 120 denier
Launderable, hand-specific gloves are sewn in a full fashion with set-in thumb. Gloves provide dexterity for protecting equipment from human contamination in assembly, examination, and inspection areas.
Specifications
- Catalog No:
- 32932-270
- 32932-272
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous