Whirl-Pak® Bags for Seward Stomacher® Lab Blenders, Nasco®
Fournisseur: NASCO SAMPLING LLC
Certificats
A propos de cet article
Sterile Whirl-Pak™ bags are designed to keep liquid, semi-solid, or solid samples in the bag during blending.
- All bags have a continuous perimeter seal
Bags are made of heavy-duty 4 mil-thick polyethylene, except for 11216-796, which is made of heavy-duty 3 mil-thick polyethylene. Bags feature patented Puncture Proof Tabs with tape extending past the wire ends to eliminate sharp points that could puncture the bag or cause damage to skin or gloves.
Ordering information: Selected styles incorporate a white, write-on strip for labeling. Sizes available for the Seward Stomacher® 400C, 3500 Series, and 80 Biomaster.
Customers Who Bought This Also Bought