A propos de cet article
Keep bags open and securely supported on these economical, lightweight, stainless steel wire frames.
- Stainless steel wire frame construction
- Autoclavable
- Economical
- Lightweight
- Polypropylene containment tray bottom
Polypropylene bottom serves as a containment tray in the event of bag leakage and is safety orange in color for easy identification as a biohazard containment system.
Dishwasher-safe and autoclave-safe, the reusable Clavies® Biohazard Bag Holder Cover opens quickly and easily by pressing the built-in release tab -no batteries required! The orange color cover fits both the Medium and Large sizes of the Clavies® Biohazard Bag Holders and features a smooth 10.1×10.1 cm (4×4") area on top for applying labels. The cover can be installed in seconds and improves the sanitation of these biohazard waste containers. The center panel of the lid opens to an upright position and reveals a large 20.3×24.1 cm (8×91⁄2") opening through which wastes can be deposited into the biohazard bags.
The small bag holder is ideal for bench top use. Assembly takes only minutes and is permanent. Dishwasher-safe and autoclavable at 121 °C (250 °F). See F13192-0102 (below) for reusable, sanitary cover to fit medium and large Clavies® Biohazard Bag Holders.