Spectra® Fiber Cut-Resistant Glove Liners, Wells Lamont
Fournisseur: Wells Lamont
Certificats
A propos de cet article
An ultrahigh molecular weight polyethylene fiber provides moderate cut resistance with high reusability.
- Gloves can be used outside
An ultrahigh molecular weight polyethylene fiber provides moderate cut resistance with high reusability.
Gloves are lightweight and have a low coefficient of friction, which allows sharp objects to slide across the surface.
Spécifications
- Catalog No:
- 32935-487
- 32935-509
- 32935-483
- 32935-485
- 32935-481
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous