VWR® Traceable® Infrared Thermometer with Pocket Clip
Catalogue # 36934-174
Fournisseur: VWR International
Spécifications
- Description:Infrared Thermometer with Pocket Clip
- Distance to Spot Ratio:1:1
- Cat. no.:36934-174
- Range:–33...220 °C (–27...428 °F)
Spécifications
À propos de cet article
Noninvasive, no-touch measurements are ideal for food preparation, life sciences, pharmaceuticals, petroleum products, cleanrooms, electronics, and field use.
- Traceable®
- Simple point-and-shoot operation
- Pocket-sized
Unit takes readings in less than a second
Field of view is 1:1; thermometer can read a spot the size of 5 cm (2") from 5 cm (2") away. The fixed emissivity of 0.95 covers most applications. Features a handy pocket clip, key-chain, and lanyard. Display indicates low battery.
Unit takes both Celsius and Fahrenheit readings of solids, semi-solids, and liquids. Hold freezes the display at the current reading.
Certifications: A serial-numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with button cell battery.