Thermo Scientific™ Serological Pipettes
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
Bold, clear bi-directional graduations make it easy-to-read fluid volumes and dispensing amounts. Choose from a broad selection of sizes and packaging options.
- Material - Ultra clear, 100% virgin polystyrene for maximum clarity
- Contaminant free - RNase/DNase free, human DNA free, non-cytotoxic, non-pyrogenic
- Sterility - E-beam irradiation to SAL of 10⁻⁶
- Shelf life - Five years from manufacturing date
Wide range of volumes and packaging options. Color-coded pipettes and packaging for convenient volume selection. Individually packed pipettes in either paper/plastic or plastic peel for high sterility assurance. Reduce packaging waste with bulk packed options. Ensures dispensing accuracy +/−2% for 1 and 2 ml and +/−1% for 5, 10, 25, and 50 ml at full volume.
Packaging: Individually wrapped, paper/plastic peel packaging.
Customers Who Bought This Also Bought