AlphaTec® Butyl 38-514 Chemical resistant gloves, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
These gloves deliver superior resistance to esters, ketones, and oxidizing agents, including concentrated oxidizing mineral acids such as 70% nitric acid and 98% sulfuric acid.
- Hand specific
- Beaded cuffs
- Superior resistance to chemicals
Ideal for hazmat, first response, unknown exposure, sampling, production, and maintenance applications. Each pair is packed with an insert containing critical information on appropriate usage.
Spécifications
- Catalog No:
- 89011-858
- 89011-860
- 89011-854
- 89011-856
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable