A propos de cet article
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Type: Secondary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: PE
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 77024-238
- 77024-450
- 77022-298
- Antigen symbol:
- CD2
- CD2
- CD2
- Antigen name:
- CD2 antigen
- CD2 antigen
- CD2 antigen
- Antibody type:
- Secondary
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- RPA-2.10
- RPA-2.10
- RPA-2.10
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse