KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Calibrated to contain
Designed from ASTM Specification E288, Class A serialized requirements. Flasks meet requirements of NBS Circular 602 and come with a Certificate of Graduation Accuracy.
Flasks are marked with individual serial numbers. Flasks have a graduation ring blasted on neck, a marking spot, and a [ST] ground glass stopper.
Customers Who Bought This Also Bought