KIMAX® Glass Rods, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Made of KG-33 borosilicate glass; which has a low coefficient of expansion
Ends are cut.
Certifications: Meets requirements for Type I Class A, ASTM E438.
Ordering information: All values are approximate.
Customers Who Bought This Also Bought