A propos de cet article
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Type: Secondary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 790
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 77021-250
- 77021-678
- Antigen symbol:
- CD19
- CD19
- Antigen name:
- CD19 antigen
- CD19 antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- SJ25C1
- SJ25C1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse