To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:CD5 antigen
- Clonality:Monoclonal
- Clone:UCHT2
- Host:Mouse
- Reactivity:Human
- Antigen symbol:CD5
- Conjugation:Unconjugated
- Size:100 µg
- Storage temperature:Refrigerated (2...8 °C); minimize light exposure
- Shipping temperature:Blue ice
- Purification:Affinity purification
- Packaging:Vial
Spécifications
A propos de cet article
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human