Wallcur® Practi-Ipratropium Bromide Nebules
# de catalogue 470355-072
Fournisseur: WALLCUR, LLC.
New Product
Spécifications
- Description:Practi-ipratropium bromide 0.02% nebules. (0.5 mg/2.5 ml)
- Cat. no.:470355-072
- Includes:Distilled water
Spécifications
A propos de cet article
Made for training.
- Contains no medication
- Contents are safe for use with most simulators
Practi-Ipratropium Bromide™ 0.02% nebules simulates 0.5 mg/2.5 ml of ipratropium bromide 0.02% (Atrovent®) in a nebulizer. Practi-ipratropium comes as a liquid solution that is to be inhaled by the patient's mouth using a special nebulizer. Safely teach this skill using Wallcur's Practi-ipratropium bromide 0.02% with any task trainer or manikin that accepts distilled water. Students can also practice dosage calculations to understand the different dosages for adults versus pediatrics.
Delivery: These products require a release form for purchase. If your account does not have one on file, the item cannot ship until release documentation is received.
Caution: For training purposes only. Not for human or animal use.
Customers Who Bought This Also Bought