VWR® Traceable® Jumbo-Display Dial Thermometers
Fournisseur: VWR International
Certificats
A propos de cet article
Thermometers fit cuvettes, test tubes, flasks, and beakers and will read temperatures in samples, reagents, water baths, foods, gases, and semi-solids.
Thermometers fit cuvettes, test tubes, flasks, and beakers. Will read temperatures in samples, reagents, water baths, foods, gases, and semi-solids. Readings update every second. Stainless steel probe is resistant to most laboratory chemicals. Dial has 5.4cm (21/8") diameter and features a large, bright 1cm (3/8") high LCD.
- Traceable®
- Wide temperature range
- Easy-to-read LCD
- Read °C and °F
Readings update every second. Stainless steel probe is resistant to most laboratory chemicals. Dial has 5.4 cm (21/8") diameter and features a large, bright 1 cm (3/8") high LCD.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with replaceable 1.5V silver-oxide battery.
Spécifications
- Catalog No:
- 61161-310
- 77776-720
- Temperature range:
- –50 to 150°C (–58 to 302°F)
- –50 to 150°C (–58 to 302°F)
- Temperature resolution:
- 0.1° from –20 to 200°, 1° outside this range
- 0.1° from –20 to 200°, 1° outside this range
- Probe:
- 133 × 3.2 mm (51/4 × 1/8")
- 133 × 3.2 mm (51/4 × 1/8")