Esteem® CP Sterile Synthetic Gloves, Cardinal
Fournisseur: Cardinal Health
Certificats
A propos de cet article
Hand-specific, powder-free clean process gloves are made of polyisoprene synthetic polymer for a fit, feel, and performance that is virtually indistinguishable from latex.
- Superior protection against bacteria and particulates
- Highly breathable and fluid-resistant
- Low particle shedding
- Soft and hypoallergenic
- Malleable noseband ensures custom, secure seal
These gloves contain no natural rubber latex proteins or allergens. Beaded cuff. Bisque finish. Sterilized by gamma radiation to a Sterility Assurance Level (SAL) of 10-6. Color: white.
Certifications: CE marked.
Ordering information: Individually pair packed, 50 pair/double poly bag, 200 pair/case.
Spécifications
- Catalog No:
- CAPH2Y72PL55
- CAPH2Y72PL60
- CAPH2Y72PL75
- CAPH2Y72PL70
- CAPH2Y72PL85
- CAPH2Y72PL80
- PH2Y72PL90
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Finger thickness:
- 11.8 mil
- 11.8 mil
- 11.8 mil
- 11.8 mil
- 11.8 mil
- 11.8 mil
- 11.8 mil
- Palm thickness:
- 10.6 mil
- 10.6 mil
- 10.6 mil
- 10.6 mil
- 10.6 mil
- 10.6 mil
- 10.6 mil