VWR® ESD Cleanroom Polyester Wipes
Supplier: VWR International
New Product
Certificates
About this item
Polyester cleanroom wipers are constructed of 100% continuous filament and double-knit polyester. To prevent the release of fibers and particles, four edges of the wipers are laser sealed .
- For use in most critical cleanroom applications
- 100% continuous filament polyester wiper
- Edges sealed with patented laser-cutting technology
- Exceptional absorbency and cleaning efficiency
- Laundered with 18MΩ DI water
- Low fiber and particulate release
The VWR® ESD Cleanroom wiper is a 100% continuous filament polyester wiper, featuring a double-knit construction. The wipe edges are sealed with laser cutting technology, resulting in very low fiber and particle release for the most critical cleanroom applications.
Customers Who Bought This Also Bought