Armored Encapsulated Glass Thermometers
Fournisseur: Heathrow Scientific
Certificats
A propos de cet article
Our precision, serialized thermometers are encapsulated in FEP. The armor is constructed of nickel-plated brass.
- Includes FEP encapsulation and nickel-plated brass armor
- Feature safety blue fill against a white background for easy reading
- Wide variety of temperature ranges available
These are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications. In the event of breakage the FEP coating contains the broken glass and liquid fill, preventing injury or contamination. The armor protects the thermometer from rough handling and shock during use.
Customers Who Bought This Also Bought