A propos de cet article
Safety/security mirrors are constructed of lightweight, 0.32cm (1/8") thick acrylic
Full dome mirrors provide a full 360° view from any angle. Ideal for four-way intersections. Supplied with hanging wires for suspension from the ceiling. Can also be attached directly to the ceiling.
Half dome (180°) mirrors are recommended for three-way (“T”) intersections. Can be wall or ceiling mounted.
Quarter dome (90°) mirrors are recommended for two-way (“L”) intersections. Screw holes are provided for corner/ceiling mounting and spring for corner/wall mounting.
Durable mirrors are unbreakable under normal conditions. Reflective surface is vacuum coated onto the inside of the dome.