Nitri-Flex™ Nitrile-Dipped Gloves, Showa
Fournisseur: Avantor
CE Compliant
Certificats
A propos de cet article
Ideal for a variety of tasks, providing an excellent lightweight grip for wet, dry, or oily materials.
- A knitwrist cuff ensures a comfortable, snug fit
- Extremely flexible gloves allow superior dexterity
Palm-dipped in nitrile, delivering stretchable, breathable comfort with unmatched abrasion and cut resistance. Palm color is green, liner color is yellow.
Spécifications
- Catalog No:
- 47744-764
- 47744-762
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Liner material:
- Kevlar®/Lycra®
- Kevlar®/Lycra®
- Finish:
- Textured
- Textured