VWR® Heavy-Duty Absorbent Wipers
Supplier: VWR International
Certificates
About this item
VWR® Industrial absorbent wipers are used in many industries and are made from recycled materials. These wipers are designed to be durable and resistant to abrasion.
- Durable and absorbent heavy-duty wipers
- Lint-free
- Strong either wet or dry
- Flat, bulk in box
These top‑tier wipers, renowned for their exceptional cleanliness and absorbency, are designed to excel in demanding environments. Crafted with a high‑performance fiber blend, they boast unparalleled lint control, ensuring a flawless finish with minimal risk of contamination. Engineered to resist snagging on various surfaces, these wipes offer both durability and reliability.
Customers Who Bought This Also Bought