Accucore™ AQ Columns
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Separate polar compounds with improved speed and resolution using Thermo Scientific™ Accucore aQ columns.
Rugged 2.6 µm solid core particles enable fast, high efficiency separations at low backpressures. The polar endcapped C18 stationary phase provides optimal retention of polar analytes and is fully compatible with 100% aqueous mobile phases. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm ID require the 1 mm UNIGUARD Holder (10046-746), 2.1/3 mm ID require the 2/3 mm UNIGUARD Holder (10046-748) and 4/4.6 mm ID require the 4/4.6 mm UNIGUARD Holder (10046-744).
Spécifications
- Catalog No:
- 10038-798
- 10038-800
- 10038-806
- 10038-808
- 10038-810
- 10039-140
- 10039-144
- 10039-146
- 10039-148
- 10039-150
- Carbon load:
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Surface area:
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g