VWR® Pocket Dew Point/Hygrometer/Thermometer
# de catalogue 12777-834
Fournisseur: VWR International
Spécifications
- Description:Pocket Hygrometer
- Temperature accuracy:±1°C (±1.8°F)
- Temperature resolution:0.1°
- Humidity range:0 to 100% RH
- Humidity resolution:0.001
- Humidity accuracy:±3% RH mid-range, ±4% RH ends of range
- Dew point range:–20…50 °C (–4…22 °F)
- Dew point resolution:0.1°
- Dew point accuracy:±1 °C (±1.8 °F)
- Temperature range:–20 to 50°C (–4 to 122°F)
- Dimensions:16.5L x 5.1W x 1.7H (61/2 x 2 x 2/3")
- Weight:85 g (3 oz.)
- Cat. no.:12777-834
Spécifications
A propos de cet article
Pocket-size meter registers relative humidity with either dew point or temperature on a 1.9 cm (3/4") high dual-display LCD.
- Traceable®
- Compact design
The device is fitted with a rugged ABS plastic case and an electronic capacitance polymer film sensor that is unaffected by condensation. At the press of a button, the unit recalls the minimum/maximum of all readings; the hold button “freezes” the display to record a reading. Typical full-range response time is a fast 30 to 60 seconds.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with two AAA batteries.
Customers Who Bought This Also Bought