ContactUsLinkComponent
Level, GEIER & BLUHM INC
Level, GEIER & BLUHM INC
# de catalogue 36680-009
Fournisseur:  Geier & Bluhm
Small Business Enterprise
Level
Level, GEIER & BLUHM INC
# de catalogue 36680-009
Fournisseur:  Geier & Bluhm
Small Business Enterprise
Spécifications
  • Description:
    Level
  • Cat. no.:
    36680-009
  • Supplier No.:
    2-10000
Spécifications
A propos de cet article
This handy little level, in 1" diameter nickel-plated metal case, is very convenient for leveling balances, scales and other instruments in the laboratory.
Customers Who Bought This Also Bought