Level, GEIER & BLUHM INC
# de catalogue 36680-009
Fournisseur: Geier & Bluhm
Small Business Enterprise
Spécifications
- Description:Level
- Cat. no.:36680-009
- Supplier No.:2-10000
Spécifications
A propos de cet article
This handy little level, in 1" diameter nickel-plated metal case, is very convenient for leveling balances, scales and other instruments in the laboratory.
Customers Who Bought This Also Bought