KIMAX® Filtering Flasks with Detachable Plastic Sidearm, Capacity Scale, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Flasks have an autoclavable plastic sidearm designed to accept 7.9mm (5/16") I.D
Designed from ASTM Specification E1406, Type III, Class 2 requirements.
flexible tubing. Sidearm has finger grips for easy insertion or removal. Opening in sidearm is larger at flask end to hold a cotton plug securely. Graduated with white scales to indicate approximate volumes.
Customers Who Bought This Also Bought