Nalgene® Disposable Erlenmeyer Flasks with Vented Closure, PETG, Sterile, Plain Bottom, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Made of lightweight, clear PETG plastic, these flasks are ideal for shaker and suspension cell culture, as well as media preparation or storage.
- Flasks feature molded-in graduations
- Non-pyrogenic, non-cytotoxic
- Sterilized to SAL 10-6
The high-density polyethylene closure has a hydrophobic 0.2µm PTFE membrane that allows sterile air exchange. Sterile disposable flasks reduce the chance of cross contamination.
Ordering information: Individually packaged.
Customers Who Bought This Also Bought