Cell Culture Storage Bottles, Polystyrene, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Disposable polystyrene bottles for storage of media, buffers and other aqueous solutions.
- Low profile, easy grip style has sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimise the risk of contamination
- Bottles can be used with Corning® vacuum filter systems
- Sterile, certified non-pyrogenic
Spécifications
- Catalog No:
- 29442-870
- 29442-878
- 29442-874
- 28199-760
- 28199-758
- 29443-026
- 29442-882
- 28199-762
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta